Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCCGGCAAGGCCCTTACCGATC[C/T]GGTCGCTATAGTGGGTGTACTGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610513 MIM: 615776 MIM: 156790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP13A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP13A2 - ATPase 13A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CROCC - ciliary rootlet coiled-coil, rootletin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFAP2 - microfibrillar associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135247.1 | 233 | Missense Mutation | CAG,CGG | Q,R 40 | NP_001128719.1 | |
NM_001135248.1 | 233 | Missense Mutation | CAG,CGG | Q,R 40 | NP_001128720.1 | |
NM_002403.3 | 233 | Missense Mutation | CAG,CGG | Q,R 41 | NP_002394.1 | |
NM_017459.2 | 233 | Missense Mutation | CAG,CGG | Q,R 41 | NP_059453.1 | |
XM_011541476.2 | 233 | Missense Mutation | CAG,CGG | Q,R 41 | XP_011539778.1 |