Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCTAGTGATTGTCCACGTCAAG[A/G]AGGCACACACACAATTTTACCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609591 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RIT1 - Ras like without CAAX 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256820.1 | 1875 | UTR 3 | NP_001243749.1 | |||
NM_001256821.1 | 1875 | UTR 3 | NP_001243750.1 | |||
NM_006912.5 | 1875 | UTR 3 | NP_008843.1 |