Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTATTATTATTATTATTATTA[C/T]TATTATTTAGGTATATCCCCTGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604513 | ||||||||||||||||||||
Literature Links: |
CD84 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD84 - CD84 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184879.1 | 5388 | UTR 3 | NP_001171808.1 | |||
NM_001184881.1 | 5388 | UTR 3 | NP_001171810.1 | |||
NM_001184882.1 | 5388 | UTR 3 | NP_001171811.1 | |||
NM_003874.3 | 5388 | UTR 3 | NP_003865.1 | |||
XM_011510094.2 | 5388 | UTR 3 | XP_011508396.1 | |||
XM_011510095.2 | 5388 | UTR 3 | XP_011508397.1 |