Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCCAATTCTCTCTCCTAGACTC[C/G]TAGGAAACGGAAATCCAGCAGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603615 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD54L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAD54L - RAD54-like (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142548.1 | 144 | Missense Mutation | CCT,CGT | P,R 32 | NP_001136020.1 | |
NM_003579.3 | 144 | Missense Mutation | CCT,CGT | P,R 32 | NP_003570.2 | |
XM_006710975.3 | 144 | UTR 5 | XP_006711038.1 | |||
XM_011542299.2 | 144 | Intron | XP_011540601.1 | |||
XM_011542300.2 | 144 | Intron | XP_011540602.1 |