Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCGCTTGAATGTCAGGAATGCT[C/T]TCCTGAAGAGCTTCTTGGAAACTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611199 MIM: 601176 | ||||||||||||||||||||
Literature Links: |
DNTTIP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNTTIP2 - deoxynucleotidyltransferase terminal interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCLM - glutamate-cysteine ligase modifier subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308253.1 | 1065 | Silent Mutation | GAA,GAG | E,E 210 | NP_001295182.1 | |
NM_002061.3 | 1065 | Silent Mutation | GAA,GAG | E,E 232 | NP_002052.1 | |
XM_011541261.2 | 1065 | Silent Mutation | GAA,GAG | E,E 144 | XP_011539563.1 | |
XM_017001056.1 | 1065 | UTR 3 | XP_016856545.1 | |||
XM_017001057.1 | 1065 | UTR 3 | XP_016856546.1 |