Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCACGTTGAGCTCTTTATATTT[G/T]TCCCCTTTTTGGGGGGCAGTGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604658 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR137B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR137B - G protein-coupled receptor 137B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003272.3 | Intron | NP_003263.1 | ||||
XM_017002209.1 | Intron | XP_016857698.1 | ||||
XM_017002210.1 | Intron | XP_016857699.1 |