Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGAAATGACACAAGTGGCTAA[C/T]TAAGGACTTTATTTCAAACAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602758 MIM: 610568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PI4KB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PI4KB - phosphatidylinositol 4-kinase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198774.1 | 3593 | UTR 3 | NP_001185703.1 | |||
XM_005245264.3 | 3593 | UTR 3 | XP_005245321.1 | |||
XM_011509634.1 | 3593 | UTR 3 | XP_011507936.1 | |||
XM_011509635.1 | 3593 | UTR 3 | XP_011507937.1 | |||
XM_011509636.2 | 3593 | UTR 3 | XP_011507938.1 | |||
XM_011509637.2 | 3593 | UTR 3 | XP_011507939.1 | |||
XM_011509638.1 | 3593 | UTR 3 | XP_011507940.1 | |||
XM_017001486.1 | 3593 | UTR 3 | XP_016856975.1 | |||
XM_017001487.1 | 3593 | UTR 3 | XP_016856976.1 | |||
XM_017001488.1 | 3593 | UTR 3 | XP_016856977.1 | |||
XM_017001489.1 | 3593 | UTR 3 | XP_016856978.1 | |||
XM_017001490.1 | 3593 | UTR 3 | XP_016856979.1 | |||
XM_017001491.1 | 3593 | UTR 3 | XP_016856980.1 | |||
XM_017001492.1 | 3593 | UTR 3 | XP_016856981.1 |
ZNF687 - zinc finger protein 687 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304763.1 | 3593 | Intron | NP_001291692.1 | |||
NM_001304764.1 | 3593 | Intron | NP_001291693.1 | |||
NM_020832.2 | 3593 | Intron | NP_065883.1 | |||
XM_005245366.3 | 3593 | Intron | XP_005245423.1 | |||
XM_011509811.2 | 3593 | Intron | XP_011508113.1 | |||
XM_011509812.2 | 3593 | Intron | XP_011508114.1 | |||
XM_011509813.2 | 3593 | Intron | XP_011508115.1 |