Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTCATTCCAGATAGGAAGACCTC[A/G]GGTCAGCTTCAGGACATAAAGTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609859 MIM: 604748 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETNK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETNK2 - ethanolamine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297760.1 | 2460 | UTR 3 | NP_001284689.1 | |||
NM_001297761.1 | 2460 | UTR 3 | NP_001284690.1 | |||
NM_001297762.1 | 2460 | UTR 3 | NP_001284691.1 | |||
NM_018208.3 | 2460 | UTR 3 | NP_060678.2 | |||
XM_011509714.2 | 2460 | UTR 3 | XP_011508016.1 | |||
XM_011509715.1 | 2460 | UTR 3 | XP_011508017.1 | |||
XM_017001632.1 | 2460 | UTR 3 | XP_016857121.1 |
SOX13 - SRY-box 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |