Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGACAGTTGGTGCTTGCATTCTA[A/T]AACATTTTAACATTTATAAAACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610771 MIM: 180474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHD5 - chromodomain helicase DNA binding protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL22 - ribosomal protein L22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000983.3 | Intron | NP_000974.1 |