Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGGAAGAAAACCTGAAAAAGAC[A/C]CCAAAGAAGAAGATGAAAATGGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611193 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRP15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RRP15 - ribosomal RNA processing 15 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016052.3 | 87 | Silent Mutation | ACA,ACC | T,T 19 | NP_057136.2 | |
XM_011509597.2 | 87 | Silent Mutation | ACA,ACC | T,T 19 | XP_011507899.1 |