Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGTCCATCATCTCCATGGTCTC[C/T]CCAAACAGCTCCTCTGCCTCTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614994 MIM: 150310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMK1G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMK1G - calcium/calmodulin dependent protein kinase IG | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LAMB3 - laminin subunit beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000228.2 | 3495 | Silent Mutation | NP_000219.2 | |||
NM_001017402.1 | 3495 | Silent Mutation | NP_001017402.1 | |||
NM_001127641.1 | 3495 | Silent Mutation | NP_001121113.1 | |||
XM_005273124.4 | 3495 | Silent Mutation | XP_005273181.1 | |||
XM_017001272.1 | 3495 | Silent Mutation | XP_016856761.1 |
MIR4260 - microRNA 4260 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |