Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTTACTTAAGTGCCAGATAGTA[C/T]AGTGTTGCCTTTAGATGTAGGTTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613709 MIM: 607020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2D - eukaryotic translation initiation factor 2D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201478.1 | Intron | NP_001188407.1 | ||||
NM_006893.2 | Intron | NP_008824.2 | ||||
XM_005277509.3 | Intron | XP_005277566.1 | ||||
XM_006711196.3 | Intron | XP_006711259.1 | ||||
XM_011509257.2 | Intron | XP_011507559.1 | ||||
XM_011509258.2 | Intron | XP_011507560.1 | ||||
XM_017000530.1 | Intron | XP_016856019.1 | ||||
XM_017000531.1 | Intron | XP_016856020.1 | ||||
XM_017000532.1 | Intron | XP_016856021.1 |
RASSF5 - Ras association domain family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182663.3 | Intron | NP_872604.1 | ||||
NM_182664.3 | Intron | NP_872605.1 | ||||
NM_182665.3 | Intron | NP_872606.1 |