Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCAGTCTCTTTTGGATTTTACG[A/G]TTCTGTAACAAGACCTACAATTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118494 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRM3 - cholinergic receptor muscarinic 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000740.2 | Intron | NP_000731.1 | ||||
XM_005273032.3 | Intron | XP_005273089.1 | ||||
XM_011544041.2 | Intron | XP_011542343.1 | ||||
XM_011544043.2 | Intron | XP_011542345.1 | ||||
XM_011544044.2 | Intron | XP_011542346.1 | ||||
XM_011544045.2 | Intron | XP_011542347.1 | ||||
XM_011544046.2 | Intron | XP_011542348.1 | ||||
XM_011544047.2 | Intron | XP_011542349.1 | ||||
XM_017000152.1 | Intron | XP_016855641.1 | ||||
XM_017000153.1 | Intron | XP_016855642.1 | ||||
XM_017000154.1 | Intron | XP_016855643.1 | ||||
XM_017000155.1 | Intron | XP_016855644.1 | ||||
XM_017000156.1 | Intron | XP_016855645.1 | ||||
XM_017000157.1 | Intron | XP_016855646.1 | ||||
XM_017000158.1 | Intron | XP_016855647.1 | ||||
XM_017000159.1 | Intron | XP_016855648.1 | ||||
XM_017000160.1 | Intron | XP_016855649.1 | ||||
XM_017000161.1 | Intron | XP_016855650.1 | ||||
XM_017000162.1 | Intron | XP_016855651.1 | ||||
XM_017000163.1 | Intron | XP_016855652.1 |