Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACAGCAAAGATCTGCCAGCCCC[A/G]GGCCTCTACCTAGTGGCCTGGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMO4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FMO4 - flavin containing monooxygenase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002022.2 | 315 | UTR 5 | NP_002013.1 | |||
XM_005245045.3 | 315 | UTR 5 | XP_005245102.1 | |||
XM_005245046.3 | 315 | UTR 5 | XP_005245103.1 | |||
XM_005245048.3 | 315 | UTR 5 | XP_005245105.1 | |||
XM_006711243.3 | 315 | UTR 5 | XP_006711306.1 | |||
XM_011509347.2 | 315 | UTR 5 | XP_011507649.1 | |||
XM_011509348.2 | 315 | UTR 5 | XP_011507650.1 | |||
XM_011509349.2 | 315 | UTR 5 | XP_011507651.1 |