Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAAGCTCATTCATGAGTACTAT[A/G]GAAACTAACAGGTTTGATAATTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616554 MIM: 604737 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPAG17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPAG17 - sperm associated antigen 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206996.2 | Intron | NP_996879.1 | ||||
XM_006710426.3 | Intron | XP_006710489.1 | ||||
XM_006710427.3 | Intron | XP_006710490.1 | ||||
XM_006710428.3 | Intron | XP_006710491.1 | ||||
XM_006710429.3 | Intron | XP_006710492.1 | ||||
XM_006710431.3 | Intron | XP_006710494.1 | ||||
XM_011540934.1 | Intron | XP_011539236.1 | ||||
XM_011540935.2 | Intron | XP_011539237.1 | ||||
XM_011540936.2 | Intron | XP_011539238.1 | ||||
XM_011540937.2 | Intron | XP_011539239.1 | ||||
XM_011540939.2 | Intron | XP_011539241.1 | ||||
XM_011540941.2 | Intron | XP_011539243.1 | ||||
XM_011540942.2 | Intron | XP_011539244.1 |
WDR3 - WD repeat domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006784.2 | Intron | NP_006775.1 |