Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGTCCACATGCCTGCGGCTCGA[A/G]GGGAAGGTTTTCATTGGCAGCCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RD3 - retinal degeneration 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164688.1 | 3428 | UTR 3 | NP_001158160.1 | |||
NM_183059.2 | 3428 | UTR 3 | NP_898882.1 | |||
XM_017001151.1 | 3428 | UTR 3 | XP_016856640.1 |