Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACATCAGATAGATCTTACAGGT[C/G]CCAGCCTGTAGACATCTTTTACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120950 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8A - complement component 8 alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000562.2 | 19 | UTR 5 | NP_000553.1 | |||
XM_011542079.2 | 19 | UTR 5 | XP_011540381.1 | |||
XM_017002234.1 | 19 | UTR 5 | XP_016857723.1 |