Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCCGGTTAAAAAACATAAAAA[A/G]CAAACAAAAAACACAAAAAATAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610656 | ||||||||||||||||||||
Literature Links: |
KYAT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KYAT3 - kynurenine aminotransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008661.2 | Intron | NP_001008661.1 | ||||
NM_001008662.2 | Intron | NP_001008662.1 |
RBMXL1 - RNA binding motif protein, X-linked like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162536.2 | Intron | NP_001156008.1 | ||||
NM_019610.5 | Intron | NP_062556.2 |