Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGGGAGAGAGGCAAGAGTTAA[A/C]AAATACTAAAATGAAGAAAGAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610393 MIM: 608741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GON4L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GON4L - gon-4 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYT11 - synaptotagmin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152280.4 | Intron | NP_689493.3 | ||||
XM_005245014.2 | Intron | XP_005245071.1 | ||||
XM_017000759.1 | Intron | XP_016856248.1 |