Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAGTATTCTTCCTTGAGGCTCA[C/T]ATCATGTGTCCCTATCACTCTTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123885 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
S100A7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
S100A7A - S100 calcium binding protein A7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176823.3 | Intron | NP_789793.1 |
S100A8 - S100 calcium binding protein A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319196.1 | Intron | NP_001306125.1 | ||||
NM_001319197.1 | Intron | NP_001306126.1 | ||||
NM_001319198.1 | Intron | NP_001306127.1 | ||||
NM_001319201.1 | Intron | NP_001306130.1 | ||||
NM_002964.4 | Intron | NP_002955.2 |