Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCACAAATCCACAGAGTTACCT[G/T]GGGGGGCACTGGCGCTTTCCCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBPF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NBPF7 - neuroblastoma breakpoint family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001047980.2 | 1234 | Missense Mutation | AAG,CAG | K,Q 297 | NP_001041445.1 | |
XM_017001186.1 | 1234 | Intron | XP_016856675.1 |