Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCTTCTCTGGGGCCTGGGACCC[A/G]TGCGGCTGGGGGAAAGCCAACAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600210 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUNX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RUNX3 - runt related transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031680.2 | 2327 | UTR 3 | NP_001026850.1 | |||
NM_001320672.1 | 2327 | UTR 3 | NP_001307601.1 | |||
NM_004350.2 | 2327 | UTR 3 | NP_004341.1 | |||
XM_005246024.4 | 2327 | UTR 3 | XP_005246081.1 | |||
XM_011542351.1 | 2327 | UTR 3 | XP_011540653.1 | |||
XM_017002670.1 | 2327 | Intron | XP_016858159.1 |