Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGTTGAAGAAATGAAGCAGTG[A/G]TTTCTTAACAGTCCTCCCCCACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMCN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMCN1 - hemicentin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031935.2 | Intron | NP_114141.2 | ||||
XM_011510038.2 | Intron | XP_011508340.1 | ||||
XM_011510041.2 | Intron | XP_011508343.1 | ||||
XM_017002437.1 | Intron | XP_016857926.1 |