Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGGAGCCTGAAGATGGCGGGGT[C/T]ATCTGCTGGAGCCTGGGGTTCTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606891 MIM: 609251 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRLA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCRLA - Fc receptor like A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FCRLB - Fc receptor like B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002901.3 | Intron | NP_001002901.1 | ||||
NM_001288829.1 | Intron | NP_001275758.1 | ||||
NM_001288830.1 | Intron | NP_001275759.1 | ||||
NM_001288831.1 | Intron | NP_001275760.1 | ||||
NM_001288832.1 | Intron | NP_001275761.1 | ||||
NM_001320241.1 | Intron | NP_001307170.1 |