Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCACGAGCCTCATCTGCTTGCT[C/G]GACAGGGGATCCTGCTAGATGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604933 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPDL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPDL - 4-hydroxyphenylpyruvate dioxygenase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032756.2 | 1201 | Missense Mutation | CGA,GGA | R,G 309 | NP_116145.1 |
MUTYH - mutY DNA glycosylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |