Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCTTCTTTGGTTAAGCACGGAG[C/T]TGAGGTGGAGGAGAGCAGTAGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G2A - phospholipase A2 group IIA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000300.3 | 819 | UTR 3 | NP_000291.1 | |||
NM_001161727.1 | 819 | UTR 3 | NP_001155199.1 | |||
NM_001161728.1 | 819 | UTR 3 | NP_001155200.1 | |||
NM_001161729.1 | 819 | UTR 3 | NP_001155201.1 |