Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTTGTTCTTCCTCTCCTGGGCT[C/G]AATCATCTGGACAAGCCTCACTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNLR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFNLR1 - interferon lambda receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_170743.3 | 3490 | UTR 3 | NP_734464.1 | |||
NM_173064.2 | 3490 | UTR 3 | NP_775087.1 | |||
NM_173065.2 | 3490 | UTR 3 | NP_775088.1 | |||
XM_006710394.3 | 3490 | UTR 3 | XP_006710457.2 | |||
XM_017000479.1 | 3490 | UTR 3 | XP_016855968.1 |