Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATTGAGCGGCAGCCCCAAGAAG[A/G]CCTTATCGTGCGCTGTGAGGCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610962 MIM: 615531 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLMP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLMP - glycosylated lysosomal membrane protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMG5 - SMG5, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323614.1 | 602 | Intron | NP_001310543.1 | |||
NM_001323615.1 | 602 | Intron | NP_001310544.1 | |||
NM_001323616.1 | 602 | Intron | NP_001310545.1 | |||
NM_001323617.1 | 602 | Intron | NP_001310546.1 | |||
NM_015327.2 | 602 | Intron | NP_056142.2 | |||
XM_017000842.1 | 602 | Intron | XP_016856331.1 | |||
XM_017000843.1 | 602 | Intron | XP_016856332.1 |
TMEM79 - transmembrane protein 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032323.2 | 602 | Missense Mutation | GAC,GGC | D,G 144 | NP_115699.1 |