Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTCTGCACTGCCCTTCTTCATC[C/T]TCACCAGTGTCCTGGGTATCCTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613665 MIM: 609743 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACKR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACKR1 - atypical chemokine receptor 1 (Duffy blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122951.2 | 380 | Missense Mutation | CTC,TTC | L,F 69 | NP_001116423.1 | |
NM_002036.3 | 380 | Missense Mutation | CTC,TTC | L,F 67 | NP_002027.2 |
CADM3 - cell adhesion molecule 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CADM3-AS1 - CADM3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |