Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAAGCTCTGCGTCTGCTGCAGGC[A/G]CAGGGCGAGCTCCCGCTCACAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602784 MIM: 601882 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APITD1-CORT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APITD1-CORT - APITD1-CORT readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CORT - cortistatin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DFFA - DNA fragmentation factor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004401.2 | 984 | Missense Mutation | CGC,TGC | R,C 296 | NP_004392.1 | |
NM_213566.1 | 984 | UTR 3 | NP_998731.1 |