Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAGCTCATCTCACGAAACTTCC[A/C]GGAGCGCACCCAACAGGTGAGGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603319 MIM: 606920 | ||||||||||||||||||||
Literature Links: |
ANXA9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANXA9 - annexin A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003568.2 | 440 | Missense Mutation | CAG,CCG | Q,P 84 | NP_003559.2 | |
XM_011510058.2 | 440 | Missense Mutation | CAG,CCG | Q,P 84 | XP_011508360.1 |
CERS2 - ceramide synthase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |