Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGAGATGCCACAACACCCAGCTG[A/G]TTTTTTTTAGAAGTTTTTGTAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601007 MIM: 613461 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LEPR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LEPR - leptin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003679.3 | Intron | NP_001003679.1 | ||||
NM_001003680.3 | Intron | NP_001003680.1 | ||||
NM_001198687.1 | Intron | NP_001185616.1 | ||||
NM_001198688.1 | Intron | NP_001185617.1 | ||||
NM_001198689.1 | Intron | NP_001185618.1 | ||||
NM_002303.5 | Intron | NP_002294.2 |
LEPROT - leptin receptor overlapping transcript | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198681.1 | Intron | NP_001185610.1 | ||||
NM_017526.4 | Intron | NP_059996.1 |