Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCACTCTGTACCTTAGTCTTTTC[A/G]GTGATAGCAAAAGCCCAGCCTCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VANGL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VANGL2 - VANGL planar cell polarity protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020335.2 | Intron | NP_065068.1 | ||||
XM_005245357.1 | Intron | XP_005245414.1 | ||||
XM_011509804.1 | Intron | XP_011508106.1 | ||||
XM_011509805.2 | Intron | XP_011508107.1 |