Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGGGACTGGGCCCGGGCGCAGA[A/C]CCAGGCCCAGGCCCAGGCCCACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRELP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRELP - proline and arginine rich end leucine rich repeat protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002725.3 | 306 | Missense Mutation | ACC,CCC | T,P 36 | NP_002716.1 | |
NM_201348.1 | 306 | Missense Mutation | ACC,CCC | T,P 36 | NP_958505.1 |