Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCACCATGAAGCTGGGCTGTGTC[C/T]TCATGGCCTGGGCCCTCTACCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606891 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRLA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCRLA - Fc receptor like A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184866.1 | 312 | Missense Mutation | CTC,TTC | L,F 24 | NP_001171795.1 | |
NM_001184867.1 | 312 | Missense Mutation | CTC,TTC | L,F 24 | NP_001171796.1 | |
NM_001184870.1 | 312 | Missense Mutation | CTC,TTC | L,F 24 | NP_001171799.1 | |
NM_001184871.1 | 312 | Missense Mutation | CTC,TTC | L,F 24 | NP_001171800.1 | |
NM_001184872.1 | 312 | Missense Mutation | CTC,TTC | L,F 24 | NP_001171801.1 | |
NM_001184873.1 | 312 | Missense Mutation | CTC,TTC | L,F 24 | NP_001171802.1 | |
NM_032738.3 | 312 | Missense Mutation | CTC,TTC | L,F 24 | NP_116127.3 | |
XM_006711581.3 | 312 | Missense Mutation | CTC,TTC | L,F 24 | XP_006711644.1 | |
XM_011510064.1 | 312 | Missense Mutation | CTC,TTC | L,F 7 | XP_011508366.1 | |
XM_011510065.1 | 312 | Missense Mutation | CTC,TTC | L,F 24 | XP_011508367.1 | |
XM_011510066.1 | 312 | Missense Mutation | CTC,TTC | L,F 24 | XP_011508368.1 |