Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGGGGTGAGGCCCGCCAAGAG[C/G]GGCTGAGTGGAGTCACAGCGGAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 138971 MIM: 611979 | ||||||||||||||||||||
Literature Links: |
CSF3R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSF3R - colony stimulating factor 3 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000760.3 | 2552 | Silent Mutation | CCC,CCG | P,P 775 | NP_000751.1 | |
NM_156039.3 | 2552 | Silent Mutation | CCC,CCG | P,P 802 | NP_724781.1 | |
NM_172313.2 | 2552 | Intron | NP_758519.1 | |||
XM_005270493.1 | 2552 | Silent Mutation | CCC,CCG | P,P 774 | XP_005270550.1 | |
XM_011540748.2 | 2552 | Silent Mutation | CCC,CCG | P,P 802 | XP_011539050.1 | |
XM_011540749.1 | 2552 | Silent Mutation | CCC,CCG | P,P 801 | XP_011539051.1 | |
XM_011540750.1 | 2552 | Silent Mutation | CCC,CCG | P,P 578 | XP_011539052.1 | |
XM_017000370.1 | 2552 | Silent Mutation | CCC,CCG | P,P 802 | XP_016855859.1 |
MRPS15 - mitochondrial ribosomal protein S15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |