Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGCGTAGCCCATCAGGGCACCA[A/G]TATTACCTATGACGACGCTGCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 147572 | ||||||||||||||||||||
Literature Links: |
IFI6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFI6 - interferon alpha inducible protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002038.3 | 467 | Missense Mutation | ACT,ATT | T,I 111 | NP_002029.3 | |
NM_022872.2 | 467 | Missense Mutation | ACT,ATT | T,I 115 | NP_075010.1 | |
NM_022873.2 | 467 | Missense Mutation | ACT,ATT | T,I 119 | NP_075011.1 |