Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGATCGAGGACGCCGAGAAGTA[C/T]TCCTTCATGGCCACCGTCACCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607826 | ||||||||||||||||||||
Literature Links: |
AHCYL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AHCYL1 - adenosylhomocysteinase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242673.1 | 373 | UTR 5 | NP_001229602.1 | |||
NM_001242674.1 | 373 | Intron | NP_001229603.1 | |||
NM_001242675.1 | 373 | Intron | NP_001229604.1 | |||
NM_001242676.1 | 373 | Intron | NP_001229605.1 | |||
NM_006621.5 | 373 | Silent Mutation | TAC,TAT | Y,Y 28 | NP_006612.2 | |
XM_011540535.1 | 373 | Silent Mutation | TAC,TAT | Y,Y 28 | XP_011538837.1 |