Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCACACAGGGCTGAGAATGCAC[C/T]GGGTATGCAAATAAATATATGACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK5 - adenylate kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012093.3 | Intron | NP_036225.2 | ||||
NM_174858.2 | Intron | NP_777283.1 | ||||
XM_005270739.4 | Intron | XP_005270796.1 | ||||
XM_006710572.3 | Intron | XP_006710635.1 | ||||
XM_017001008.1 | Intron | XP_016856497.1 | ||||
XM_017001009.1 | Intron | XP_016856498.1 | ||||
XM_017001010.1 | Intron | XP_016856499.1 | ||||
XM_017001011.1 | Intron | XP_016856500.1 | ||||
XM_017001012.1 | Intron | XP_016856501.1 |