Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTTCTGGTGGCCGGGACGCTCT[C/G]CTTCGCTTGGTGGAGCGAAGGGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM61 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM61 - transmembrane protein 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182532.2 | 503 | Missense Mutation | TCC,TGC | S,C 34 | NP_872338.1 | |
XM_005270586.4 | 503 | Missense Mutation | TCC,TGC | S,C 29 | XP_005270643.1 | |
XM_011540911.2 | 503 | Missense Mutation | TCC,TGC | S,C 34 | XP_011539213.1 | |
XM_011540912.2 | 503 | Missense Mutation | TCC,TGC | S,C 29 | XP_011539214.1 |