Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATAAATTTGGTTTCAAAAATAC[A/G]GAGTTGGGGGTTAATGGGACATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608057 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNASE2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNASE2B - deoxyribonuclease II beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021233.2 | Intron | NP_067056.2 | ||||
NM_058248.1 | Intron | NP_490649.1 | ||||
XM_011541878.2 | Intron | XP_011540180.1 |