Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCAGCTCTTCATCACCTCCAA[C/G]GTATCTTGGTTAAGTTGGCTGATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607567 | ||||||||||||||||||||
Literature Links: |
OLFM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OLFM3 - olfactomedin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288821.1 | 1396 | Silent Mutation | ACC,ACG | T,T 378 | NP_001275750.1 | |
NM_001288823.1 | 1396 | Silent Mutation | ACC,ACG | T,T 283 | NP_001275752.1 | |
NM_058170.3 | 1396 | Silent Mutation | ACC,ACG | T,T 358 | NP_477518.2 | |
XM_017000240.1 | 1396 | Silent Mutation | ACC,ACG | T,T 283 | XP_016855729.1 |