Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTGGGCAGTGCTCATATTGTGT[C/G]AGGTCATACACTCGTACCTGGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610742 MIM: 609957 MIM: 165380 | ||||||||||||||||||||
Literature Links: |
FAM19A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM19A3 - family with sequence similarity 19 member A3, C-C motif chemokine like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105378910 - uncharacterized LOC105378910 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOV10 - Mov10 RISC complex RNA helicase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPM1J - protein phosphatase, Mg2+/Mn2+ dependent 1J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005167.5 | 1263 | Silent Mutation | CTC,CTG | L,L 412 | NP_005158.5 |
RHOC - ras homolog family member C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |