Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTAATTGGGGCAAACTTCTCAAT[C/T]ACAGCATCTGCCACGGCCAGCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WARS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WARS2 - tryptophanyl tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015836.3 | 780 | Silent Mutation | GTA,GTG | V,V 310 | NP_056651.1 | |
NM_201263.2 | 780 | UTR 3 | NP_957715.1 | |||
XM_005270350.3 | 780 | Silent Mutation | GTA,GTG | V,V 292 | XP_005270407.1 | |
XM_011540494.2 | 780 | Silent Mutation | GTA,GTG | V,V 287 | XP_011538796.1 | |
XM_011540495.2 | 780 | Silent Mutation | GTA,GTG | V,V 224 | XP_011538797.1 | |
XM_017000038.1 | 780 | Silent Mutation | GTA,GTG | V,V 291 | XP_016855527.1 | |
XM_017000039.1 | 780 | Silent Mutation | GTA,GTG | V,V 287 | XP_016855528.1 | |
XM_017000040.1 | 780 | Silent Mutation | GTA,GTG | V,V 253 | XP_016855529.1 | |
XM_017000041.1 | 780 | Silent Mutation | GTA,GTG | V,V 197 | XP_016855530.1 | |
XM_017000042.1 | 780 | UTR 3 | XP_016855531.1 |