Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGGAGAAAGCCACGGAACAGAT[G/T]ATCTCCCTGCAGTGGACATGTCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606897 | ||||||||||||||||||||
Literature Links: |
LYST PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LYST - lysosomal trafficking regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000081.3 | 11731 | Silent Mutation | ATA,ATC | I,I 3704 | NP_000072.2 | |
NM_001301365.1 | 11731 | Silent Mutation | ATA,ATC | I,I 3704 | NP_001288294.1 | |
XM_011544031.1 | 11731 | Silent Mutation | ATA,ATC | I,I 3758 | XP_011542333.1 | |
XM_011544032.1 | 11731 | Silent Mutation | ATA,ATC | I,I 3758 | XP_011542334.1 | |
XM_011544033.2 | 11731 | Silent Mutation | ATA,ATC | I,I 3758 | XP_011542335.1 | |
XM_011544034.1 | 11731 | Silent Mutation | ATA,ATC | I,I 3712 | XP_011542336.1 | |
XM_011544035.2 | 11731 | Intron | XP_011542337.1 | |||
XM_011544036.2 | 11731 | Silent Mutation | ATA,ATC | I,I 2979 | XP_011542338.1 | |
XM_011544037.2 | 11731 | Intron | XP_011542339.1 | |||
XM_011544039.2 | 11731 | Intron | XP_011542341.1 | |||
XM_011544040.2 | 11731 | Intron | XP_011542342.1 | |||
XM_017000150.1 | 11731 | Silent Mutation | ATA,ATC | I,I 3681 | XP_016855639.1 | |
XM_017000151.1 | 11731 | Intron | XP_016855640.1 |