Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGTCAACCCAGGAACTGAGGGA[A/G]TGAGGGAACCCTGGGGAATTCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147485 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IPP - intracisternal A particle-promoted polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145349.1 | Intron | NP_001138821.1 | ||||
NM_005897.2 | Intron | NP_005888.1 | ||||
XM_005270839.4 | Intron | XP_005270896.1 | ||||
XM_006710623.3 | Intron | XP_006710686.1 | ||||
XM_011541394.2 | Intron | XP_011539696.1 | ||||
XM_017001210.1 | Intron | XP_016856699.1 |
TMEM69 - transmembrane protein 69 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |