Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATACCAGGTGTAGTTTACAGACTC[A/G]CCAGGTATCACACATGACAGTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD48 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD48 - CD48 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256030.1 | 630 | Silent Mutation | GGC,GGT | G,G 158 | NP_001242959.1 | |
NM_001778.3 | 630 | Silent Mutation | GGC,GGT | G,G 158 | NP_001769.2 | |
XM_005245625.1 | 630 | Intron | XP_005245682.1 | |||
XM_011510171.1 | 630 | Silent Mutation | GGC,GGT | G,G 126 | XP_011508473.1 | |
XM_017002867.1 | 630 | Silent Mutation | GGC,GGT | G,G 57 | XP_016858356.1 |