Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACTTGATGGAGGAAAAGTTCCC[A/G]GGTGATGCCGGTTTGGGCAAACTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PYHIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PYHIN1 - pyrin and HIN domain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152501.4 | 421 | Silent Mutation | CCA,CCG | P,P 57 | NP_689714.2 | |
NM_198928.4 | 421 | Silent Mutation | CCA,CCG | P,P 57 | NP_945146.1 | |
NM_198929.4 | 421 | Silent Mutation | CCA,CCG | P,P 57 | NP_945147.1 | |
NM_198930.3 | 421 | Silent Mutation | CCA,CCG | P,P 57 | NP_945148.1 | |
XM_005244930.1 | 421 | Silent Mutation | CCA,CCG | P,P 57 | XP_005244987.1 | |
XM_011509242.2 | 421 | Silent Mutation | CCA,CCG | P,P 57 | XP_011507544.1 | |
XM_011509243.2 | 421 | Silent Mutation | CCA,CCG | P,P 57 | XP_011507545.1 | |
XM_017000463.1 | 421 | Silent Mutation | CCA,CCG | P,P 57 | XP_016855952.1 |