Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACAGGCAGAGGCCCCGCAGAGC[A/G]CGGGTGGAGGTGGGAGCAGGCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605097 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC45A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC45A3 - solute carrier family 45 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033102.2 | 1881 | Missense Mutation | GCG,GTG | A,V 459 | NP_149093.1 | |
XM_005245556.3 | 1881 | Missense Mutation | GCG,GTG | A,V 459 | XP_005245613.1 | |
XM_005245557.4 | 1881 | Missense Mutation | GCG,GTG | A,V 459 | XP_005245614.1 | |
XM_005245560.2 | 1881 | Intron | XP_005245617.1 | |||
XM_017002638.1 | 1881 | Missense Mutation | GCG,GTG | A,V 459 | XP_016858127.1 |