Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGCGGGCAGGACACGGAGGAC[C/T]GGAGCGGCCTGTCCGAGGTGAGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SFT2D2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SFT2D2 - SFT2 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199344.2 | 109 | Missense Mutation | CGG,TGG | R,W 16 | NP_955376.1 |